Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029639.2 | NP_083915.2 | 1, 2 | 222 | ||
GenBank mRNA | AK002767.1 | - | 1, 2 | 224 | ||
AK005558.1 | - | 1, 2 | 223 | |||
AY364436.1 | - | 1, 2 | 222 | |||
BC017624.1 | - | 1, 2 | 206 | 4487871 | ||
BC022950.1 | - | 1, 2 | 202 | 5148408 |