Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Styxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289554.1 | NP_001276483.1 | 6, 7 | 759 | ||
NM_001289555.1 | NP_001276484.1 | 6, 7 | 756 | |||
NM_001289556.1 | NP_001276485.1 | 5, 6 | 600 | |||
NM_029659.4 | NP_083935.3 | 6, 7 | 766 | |||
GenBank mRNA | AK005599.1 | - | 6, 7 | 757 | ||
AK005858.1 | - | 5, 6 | 507 | |||
AK161310.1 | - | 6, 7 | 767 | |||
BC045148.1 | - | 6, 7 | 696 | 6307739 |