Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spatc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029661.1 | NP_083937.1 | 2 | 580 | ||
GenBank mRNA | AK006410.1 | - | 2 | 580 | ||
BC049686.1 | - | 2 | 571 | |||
BC147585.1 | - | 2 | 560 | 9056575 | ||
BC147614.1 | - | 2 | 560 | 9056604 |