Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029688.5 | NP_083964.2 | 1 | 341 | ||
GenBank mRNA | AK007296.1 | - | 1 | 136 | ||
AK089976.1 | - | 1 | 138 | |||
AK159226.1 | - | 1 | 136 | |||
AK159613.1 | - | 1 | 135 | |||
AK161066.1 | - | 1 | 178 | |||
BC011325.1 | - | 1 | 155 | 3154456 | ||
BC049957.1 | - | 1 | 341 | 4507800 |