Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdh1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029696.4 | NP_083972.3 | 6, 7 | 1071 | ||
GenBank mRNA | AK007262.1 | - | 2, 3 | 196 | ||
BC050786.1 | - | 6, 7 | 1071 | 6703279 |