Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165991.1 | NP_001159463.1 | 8 | 1506 | ||
NM_001165992.1 | NP_001159464.1 | 8 | 1487 | |||
NM_001165995.1 | NP_001159467.1 | 7 | 1393 | |||
NM_029702.4 | NP_083978.3 | 8 | 1500 | |||
GenBank mRNA | AK032592.1 | - | 2 | 1284 | ||
AK044759.1 | - | 8 | 1458 | |||
BC046782.1 | - | 7 | 1367 | 5720540 | ||
BC046786.1 | - | 7 | 1367 | 6416321 |