Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029738.2 | NP_084014.1 | 2 | 219 | ||
GenBank mRNA | AK009540.1 | - | 2 | 190 | ||
AK011846.1 | - | 3 | 247 | |||
AK134648.1 | - | 2 | 186 | |||
AK166050.1 | - | 2 | 300 | |||
BC024910.1 | - | 2 | 219 | 3603454 |