Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB095735.1 | - | 2 | 233 | ||
AB095736.1 | - | 2 | 233 | |||
AK012919.1 | - | 2 | 234 | |||
AK054412.1 | - | 2 | 223 | |||
AK133177.1 | - | 2 | 232 | |||
AK154594.1 | - | 2 | 241 | |||
AK164249.1 | - | 2 | 232 | |||
AK164999.1 | - | 2 | 239 | |||
AK166351.1 | - | 2 | 241 | |||
AK169601.1 | - | 2 | 232 | |||
BB841951.1 | - | 2 | 262 | |||
BC060208.1 | - | 2 | 193 | |||
CF898098.1 | - | 2 | 255 |