Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029789.1 | NP_084065.1 | 4 | 487 | ||
GenBank mRNA | AK002626.1 | - | 4 | 455 | ||
AK050161.1 | - | 4 | 468 | |||
AK146106.1 | - | 4 | 466 | |||
AK159325.1 | - | 4 | 466 | |||
AK216148.1 | - | 1 | 22 | |||
AY029532.1 | - | 4 | 487 | |||
BC006847.1 | - | 4 | 459 | 3598473 |