Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290649.1 | NP_001277578.1 | 12 | 1342 | ||
NM_029793.2 | NP_084069.1 | 13 | 1417 | |||
GenBank mRNA | AK028516.1 | - | 13 | 1337 | ||
AK044987.1 | - | 12 | 1827 | |||
BC037037.1 | - | 8 | 753 | |||
BC043090.1 | - | 13 | 1282 | 6418353 | ||
BC053068.1 | - | 12 | 1270 | 6405595 | ||
BC080733.1 | - | 12 | 1342 | 30110235 |