Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164363.1 | NP_001157835.1 | 4 | 732 | ||
NM_001164365.1 | NP_001157837.1 | 4 | 389 | |||
NM_029810.4 | NP_084086.3 | 4 | 320 | |||
GenBank mRNA | AK008045.1 | - | 4 | 267 | ||
AK030786.1 | - | 5 | 337 | |||
AK036730.1 | - | 4 | 278 | |||
AK038458.1 | - | 1 | 25 | |||
AK132384.1 | - | 4 | 278 | |||
AK147901.1 | - | 4 | 278 | |||
AK162385.1 | - | 4 | 697 | |||
AK169616.1 | - | 4 | 278 | |||
AK181552.1 | - | 3 | 181 | |||
AK182698.1 | - | 2 | 65 | |||
AK187621.1 | - | 3 | 181 | |||
AK201286.1 | - | 2 | 122 | |||
AK211414.1 | - | 2 | 122 | |||
BC064760.1 | - | 3 | 254 | 30436992 |