Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029814.1 | NP_084090.1 | 8 | 802 | ||
GenBank mRNA | AK003313.1 | - | 3 | 551 | ||
AK008911.1 | - | 8 | 802 | |||
AK147058.1 | - | 8 | 824 | |||
BC006947.1 | - | 8 | 763 | 2811290 | ||
BC080673.1 | - | 7 | 1597 |