Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164369.1 | NP_001157841.1 | 4 | 680 | ||
NM_029815.2 | NP_084091.2 | 4 | 680 | |||
GenBank mRNA | AK008957.1 | - | 1 | 86 | ||
AK020254.1 | - | 4 | 681 | |||
AK031825.1 | - | 1 | 86 | |||
AY219233.1 | - | 4 | 663 | |||
BC113166.1 | - | 4 | 660 | 40059981 | ||
BC113957.1 | - | 4 | 660 | 40059979 |