Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164369.1 | NP_001157841.1 | 12 | 2021 | ||
NM_029815.2 | NP_084091.2 | 13 | 2159 | |||
GenBank mRNA | AK008957.1 | - | 10 | 1565 | ||
AK031825.1 | - | 7 | 1292 | |||
AY219233.1 | - | 13 | 2142 | |||
BC024553.1 | - | 3 | 572 | |||
BC113166.1 | - | 12 | 2004 | 40059981 | ||
BC113957.1 | - | 12 | 2001 | 40059979 |