Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205391.1 | NP_001192320.1 | 10 | 1130 | ||
NM_001205392.1 | NP_001192321.1 | 8 | 1025 | |||
NM_029846.4 | NP_084122.2 | 9 | 1082 | |||
GenBank mRNA | AB087879.1 | - | 8 | 836 | ||
AB087880.1 | - | 9 | 893 | |||
AB087881.1 | - | 10 | 941 | |||
AB476646.1 | - | 10 | 941 | |||
AK005181.1 | - | 10 | 1055 | |||
AK030983.1 | - | 9 | 1023 | |||
AK131120.1 | - | 8 | 1868 | |||
AK155358.1 | - | 8 | 961 | |||
AK163246.1 | - | 8 | 964 | |||
AK170073.1 | - | 8 | 959 | |||
AK218161.1 | - | 4 | 288 | |||
BC049122.1 | - | 10 | 1055 | 6813377 |