Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205391.1 | NP_001192320.1 | 13 | 1380 | ||
NM_001205392.1 | NP_001192321.1 | 11 | 1275 | |||
NM_029846.4 | NP_084122.2 | 12 | 1332 | |||
GenBank mRNA | AB087879.1 | - | 11 | 1086 | ||
AB087880.1 | - | 12 | 1143 | |||
AB087881.1 | - | 13 | 1191 | |||
AB476646.1 | - | 13 | 1191 | |||
AK005181.1 | - | 13 | 1305 | |||
AK030983.1 | - | 12 | 1273 | |||
AK131120.1 | - | 11 | 2118 | |||
AK155358.1 | - | 11 | 1211 | |||
AK163246.1 | - | 11 | 1214 | |||
AK170073.1 | - | 11 | 1209 | |||
BC033362.1 | - | 3 | 653 | |||
BC049122.1 | - | 13 | 1305 | 6813377 |