Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029847.4 | NP_084123.2 | 4 | 638 | ||
GenBank mRNA | AK013194.2 | - | 4 | 590 | ||
AK019515.1 | - | 4 | 604 | |||
AK031147.1 | - | 4 | 626 | |||
AK032812.1 | - | 4 | 604 | |||
AK033968.1 | - | 4 | 639 | |||
AK035464.1 | - | 4 | 559 | |||
AK039765.1 | - | 4 | 634 | |||
AK046378.1 | - | 4 | 639 | |||
AK083188.1 | - | 4 | 592 | |||
AK084090.1 | - | 4 | 624 | |||
AK086667.1 | - | 4 | 597 | |||
AK139856.1 | - | 2 | 178 | |||
AK144817.1 | - | 4 | 606 | |||
AK165970.1 | - | 4 | 606 | |||
BC046790.1 | - | 4 | 612 | 5717223 | ||
BC058351.1 | - | 4 | 612 | 5717223 |