Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029847.4 | NP_084123.2 | 8 | 1561 | ||
GenBank mRNA | AK013194.2 | - | 8 | 1513 | ||
AK019515.1 | - | 8 | 1527 | |||
AK031147.1 | - | 8 | 1549 | |||
AK032812.1 | - | 8 | 1527 | |||
AK035464.1 | - | 8 | 1482 | |||
AK039765.1 | - | 8 | 1557 | |||
AK046378.1 | - | 8 | 1562 | |||
AK083188.1 | - | 8 | 1515 | |||
AK084090.1 | - | 8 | 1547 | |||
AK086667.1 | - | 8 | 1520 | |||
AK144817.1 | - | 8 | 1529 | |||
AK165970.1 | - | 8 | 1529 | |||
BC046790.1 | - | 8 | 1535 | 5717223 | ||
BC058351.1 | - | 8 | 1535 | 5717223 |