Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006097.4 | NP_006088.2 | 2 | 220 | ||
NM_181526.2 | NP_852667.1 | 2 | 220 | |||
GenBank mRNA | AF176042.1 | - | 1 | 156 | ||
AF176043.1 | - | 1 | 156 | |||
AK097235.1 | - | 1 | 3476 | |||
AK130879.1 | - | 2 | 185 | |||
AL697699.1 | - | 2 | 220 | |||
BC002648.2 | - | 2 | 159 | 3608316 | ||
BQ953325.1 | - | 210 | ||||
CR456843.1 | - | 1 | 116 | |||
D82057.1 | - | 2 | 188 | |||
J02854.1 | - | 1 | 158 |