Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168403.1 | NP_001161875.1 | 5 | 945 | ||
NM_001168404.1 | NP_001161876.1 | 5 | 482 | |||
NM_001168405.1 | NP_001161877.1 | 4 | 839 | |||
NM_029898.3 | NP_084174.2 | 4 | 839 | |||
GenBank mRNA | AK039553.1 | - | 4 | 840 | ||
AK045480.1 | - | 4 | 473 | |||
AK081197.1 | - | 5 | 483 | |||
AK138273.1 | - | 4 | 494 | |||
AK140417.1 | - | 5 | 809 | |||
AK155092.1 | - | 3 | 280 | |||
AK171119.1 | - | 3 | 280 | |||
BC152317.1 | - | 4 | 466 | 40088531 | ||
BC152536.1 | - | 4 | 466 | 40088530 |