Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029901.2 | NP_084177.2 | 2, 3 | 334 | ||
GenBank mRNA | AB178898.1 | - | 2, 3 | 260 | ||
AI882115.1 | - | 2, 3 | 332 | |||
AK020439.1 | - | 2, 3 | 334 | |||
AY742217.1 | - | 2, 3 | 260 | |||
BC013531.1 | - | 2, 3 | 310 | 4238756 | ||
BC061057.1 | - | 2, 3 | 330 | 30305353 | ||
BC091761.1 | - | 2, 3 | 310 | 5044038 | ||
BY721648.1 | - | 2, 3 | 334 |