Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029934.3 | NP_084210.2 | 8 | 2845 | ||
GenBank mRNA | AB055410.1 | - | 8 | 2843 | ||
AK150010.1 | - | 8 | 2239 | |||
AK154606.1 | - | 8 | 2844 | |||
AK158998.1 | - | 6 | 3766 | |||
AK170124.1 | - | 8 | 2322 | |||
AK183294.1 | - | 1 | 376 | |||
AK211361.1 | - | 1 | 352 | |||
BC023417.1 | - | 8 | 2293 | 5028759 |