Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029946.4 | NP_084222.4 | 4 | 366 | ||
GenBank mRNA | BC065111.1 | - | 4 | 366 | 30536704 |