Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000841.3 | NP_000832.1 | 2 | 615 | ||
NM_001256811.2 | NP_001243740.1 | 2 | 615 | |||
NM_001282847.1 | NP_001269776.1 | 2 | 615 | |||
XM_017010790.1 | XP_016866279.1 | 2 | 553 | |||
XM_017010791.1 | XP_016866280.1 | 2 | 608 | |||
GenBank mRNA | AK289524.1 | - | 933 | |||
AK289534.1 | - | 2 | 558 | |||
AK312300.1 | - | 2 | 570 | |||
BC130526.1 | - | 1 | 273 | 40146557 | ||
BC130528.1 | - | 1 | 273 | 40146559 | ||
BC144057.1 | - | 1 | 273 | 9052577 | ||
EU432124.1 | - | 1 | 114 | |||
U92457.1 | - | 1 | 139 | |||
X80818.1 | - | 1 | 284 |
Set Membership: |
G-protein coupled receptor |