Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp955a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029952.3 | NP_084228.2 | 4 | 649 | ||
GenBank mRNA | AK038946.1 | - | 4 | 650 | ||
AK146709.1 | - | 4 | 645 | |||
BC043067.1 | - | 4 | 612 | 5709349 | ||
BC094586.1 | - | 4 | 629 | 30620063 |
Gene Symbol: Zfp955b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142957.1 | NP_001136429.1 | 4 | 649 | ||
GenBank mRNA | AK021244.1 | - | 4 | 647 | ||
AK161843.1 | - | 4 | 649 |