Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029985.2 | NP_084261.1 | 2 | 401 | ||
GenBank mRNA | AK020847.1 | - | 2 | 207 | ||
AK143870.1 | - | 1 | 175 | |||
BC027203.1 | - | 2 | 333 | 3710091 | ||
BC031894.1 | - | 2 | 305 | 5362699 | ||
CJ174174.1 | - | 2 | 402 |