Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030153.2 | NP_084429.2 | 2 | 223 | ||
GenBank mRNA | AK021042.1 | - | 2 | 114 | ||
AY102701.1 | - | 2 | 145 | |||
BC027201.1 | - | 2 | 159 | |||
BC043701.1 | - | 2 | 136 | |||
BC056435.1 | - | 2 | 132 | 5707801 | ||
BY100294.1 | - | 2 | 238 |