Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc30a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030188.3 | NP_084464.3 | 1 | 694 | ||
GenBank mRNA | AK019670.1 | - | 1 | 568 | ||
AK029535.1 | - | 497 | ||||
AK045096.1 | - | 1 | 494 | |||
BC004773.1 | - | 1 | 489 | 3256753 | ||
BU610816.1 | - | 1 | 694 |
Gene Symbol: Ttc30a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081228.1 | NP_001074697.1 | 1 | 477 | ||
GenBank mRNA | BC157934.1 | - | 1 | 498 | 9088113 | |
BC158130.1 | - | 1 | 594 | 9007398 | ||
BC172090.1 | - | 1 | 498 | 9054376 |