Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030205.4 | NP_084481.3 | 2 | 245 | ||
GenBank mRNA | AK018739.1 | - | 2 | 213 | ||
AK037894.1 | - | 2 | 239 | |||
AK039585.1 | - | 2 | 302 | |||
AK040408.1 | - | 251 | ||||
AK088155.1 | - | 2 | 213 | |||
AK143624.1 | - | 2 | 204 | |||
AK150051.1 | - | 268 | ||||
AK178516.1 | - | 1 | 39 | |||
AK183197.1 | - | 1 | 22 | |||
BC061006.1 | - | 2 | 245 | 6536106 |