Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040408.1 | - | 3615 | |||
AK149073.1 | - | 498 | ||||
AK150051.1 | - | 3632 | ||||
AK160969.1 | - | 1768 |