Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030225.4 | NP_084501.1 | 15 | 1531 | ||
GenBank mRNA | AK054053.1 | - | 15 | 1485 | ||
AK149664.1 | - | 15 | 1507 | |||
AK158877.1 | - | 15 | 1532 | |||
AK168570.1 | - | 15 | 1507 | |||
AK169943.1 | - | 15 | 1485 | |||
AK212893.1 | - | 1 | 192 | |||
AK218237.1 | - | 1 | 192 | |||
BC006702.1 | - | 15 | 1468 | 3583936 |