Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030253.3 | NP_084529.1 | 4 | 1000 | ||
GenBank mRNA | AK037903.1 | - | 4 | 567 | ||
AK050032.1 | - | 4 | 444 | |||
BC003281.1 | - | 4 | 442 | 3581841 | ||
BC010312.1 | - | 3 | 425 | |||
BC070466.1 | - | 4 | 1000 | 30615225 |