Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pofut2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030262.3 | NP_084538.2 | 6 | 813 | ||
GenBank mRNA | AF455270.1 | - | 6 | 803 | ||
AK132373.1 | - | 6 | 810 | |||
AK147414.1 | - | 3 | 4671 | |||
AK148244.1 | - | 6 | 813 | |||
AK206297.1 | - | 2 | 113 | |||
BC003494.1 | - | 2 | 165 | |||
BC018194.1 | - | 6 | 811 | |||
BC147077.1 | - | 6 | 880 | 9056064 | ||
BC147078.1 | - | 6 | 880 | 9056063 |