Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199242.1 | NP_001186171.1 | 9 | 1061 | ||
NM_001199243.1 | NP_001186172.1 | 8 | 959 | |||
NM_001199244.1 | NP_001186173.1 | 8 | 1102 | |||
NM_001199245.1 | NP_001186174.1 | 8 | 955 | |||
NM_030265.3 | NP_084541.3 | 8 | 887 | |||
GenBank mRNA | AK039048.1 | - | 8 | 888 | ||
AK049662.1 | - | 8 | 839 | |||
AK133416.1 | - | 8 | 807 | |||
AK148685.1 | - | 8 | 1104 | |||
AK165773.1 | - | 8 | 829 | |||
BC051130.1 | - | 9 | 1061 | 6532082 | ||
DQ148501.1 | - | 9 | 1061 | |||
DQ148502.1 | - | 7 | 819 | |||
DQ148503.1 | - | 8 | 839 | |||
DQ148504.1 | - | 8 | 955 |