Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030559.3 | NP_085036.3 | 5 | 528 | ||
GenBank mRNA | AB056721.1 | - | 5 | 486 | ||
AK049872.1 | - | 5 | 485 | |||
AK162984.1 | - | 5 | 495 | |||
AK169694.1 | - | 4 | 591 | |||
BC025626.1 | - | 5 | 494 | 5320572 |