Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006142.3 | NP_006133.1 | 1 | 709 | ||
GenBank mRNA | AF029082.1 | - | 1 | 709 | ||
BC000329.2 | - | 1 | 689 | 2822397 | ||
BC000995.2 | - | 1 | 686 | 3445915 | ||
BC001550.1 | - | 2 | 595 | 3457665 | ||
BC002995.1 | - | 1 | 689 | 2822397 | ||
BC023552.2 | - | 1 | 713 | 3534328 | ||
CR541905.1 | - | 1 | 638 | |||
CR541926.1 | - | 1 | 638 | |||
M93010.1 | - | 1 | 648 |