Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030559.3 | NP_085036.3 | 19 | 1937 | ||
GenBank mRNA | AB056721.1 | - | 19 | 1895 | ||
AK032558.1 | - | 10 | 898 | |||
AK049872.1 | - | 19 | 1894 | |||
AK162984.1 | - | 16 | 2260 | |||
AK169694.1 | - | 14 | 2670 | |||
BC003944.1 | - | 7 | 809 | 3487257 | ||
BC025626.1 | - | 19 | 1903 | 5320572 |