Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077687.1 | NP_001071155.1 | 3 | 565 | ||
NM_001077688.1 | NP_001071156.1 | 3 | 565 | |||
NM_030695.2 | NP_109620.2 | 3 | 565 | |||
GenBank mRNA | AF187731.1 | - | 3 | 587 | ||
AF188506.1 | - | 3 | 587 | |||
AF188507.1 | - | 3 | 587 | |||
AK052148.1 | - | 3 | 752 | |||
AK085208.1 | - | 3 | 554 |