Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163348.1 | NP_001156820.1 | 7 | 2880 | ||
NM_001163349.1 | NP_001156821.1 | 6 | 2745 | |||
NM_001163350.1 | NP_001156822.1 | 8 | 2871 | |||
NM_030699.2 | NP_109624.1 | 8 | 3048 | |||
NM_133488.1 | NP_597995.1 | 7 | 2811 | |||
GenBank mRNA | AB038667.1 | - | 8 | 2825 | ||
AK034210.1 | - | 1 | 1296 | |||
AK034465.1 | - | 7 | 2345 | |||
AK134467.1 | - | 6 | 2743 | |||
AK173075.1 | - | 6 | 2530 | |||
BC079881.1 | - | 6 | 2712 | 6837530 |