Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mesdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030705.4 | NP_109630.1 | 1 | 2376 | ||
GenBank mRNA | AK036338.1 | - | 2342 | |||
AK084510.1 | - | 1 | 2322 | |||
AK162446.1 | - | 1 | 2263 | |||
AK195918.1 | - | 1 | 197 | |||
AK206284.1 | - | 1 | 196 |