Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271725.1 | NP_001258654.1 | 3 | 363 | ||
NM_001271726.1 | NP_001258655.1 | 3 | 399 | |||
NM_001271727.1 | NP_001258656.1 | 3 | 668 | |||
NM_001271728.1 | NP_001258657.1 | 3 | 466 | |||
NM_030706.3 | NP_109631.2 | 3 | 462 | |||
GenBank mRNA | AB043550.1 | - | 3 | 446 | ||
AF220017.1 | - | 3 | 311 | |||
AK039741.1 | - | 3 | 460 | |||
AK045410.1 | - | 3 | 459 | |||
AK046887.1 | - | 3 | 447 | |||
AK079415.1 | - | 3 | 613 | |||
AK083057.1 | - | 2 | 2221 | |||
AK141419.1 | - | 3 | 309 | |||
AK147275.1 | - | 3 | 576 | |||
AK147395.1 | - | 3 | 457 | |||
AK164350.1 | - | 2 | 165 | |||
AK220343.1 | - | 3 | 667 | |||
BC058668.1 | - | 3 | 430 | |||
BC058961.1 | - | 3 | 466 | 6826143 | ||
CD802800.1 | - | 3 | 325 | |||
CD803806.1 | - | 3 | 462 | |||
CJ100776.1 | - | 3 | 364 |