Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lactb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030717.1 | NP_109642.1 | 3 | 665 | ||
GenBank mRNA | AF317900.1 | - | 3 | 665 | ||
AK078550.1 | - | 3 | 606 | |||
AK149410.1 | - | 3 | 608 | |||
AK187555.1 | - | 1 | 128 | |||
BC016583.1 | - | 3 | 592 | |||
BC021604.1 | - | 3 | 591 | |||
BC046293.1 | - | 3 | 592 | 6484767 | ||
BC147809.1 | - | 3 | 610 | 9087526 |