Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lactb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030717.1 | NP_109642.1 | 6 | 1523 | ||
GenBank mRNA | AF317900.1 | - | 6 | 1523 | ||
AK078550.1 | - | 5 | 1298 | |||
BC046293.1 | - | 6 | 1450 | 6484767 | ||
BC147809.1 | - | 6 | 1468 | 9087526 |