Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lactb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030717.1 | NP_109642.1 | 4, 5 | 1062 | ||
GenBank mRNA | AF317900.1 | - | 4, 5 | 1062 | ||
AK213700.1 | - | 1, 2 | 132 | |||
BC046293.1 | - | 4, 5 | 989 | 6484767 | ||
BC147809.1 | - | 4, 5 | 1007 | 9087526 |