Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pum2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160219.1 | NP_001153691.1 | 3 | 327 | ||
NM_001160220.1 | NP_001153692.1 | 3 | 327 | |||
NM_001160221.1 | NP_001153693.1 | 3 | 142 | |||
NM_001160222.1 | NP_001153694.1 | 3 | 142 | |||
NM_030723.2 | NP_109648.2 | 3 | 251 | |||
GenBank mRNA | AF315590.1 | - | 3 | 267 | ||
AK122225.1 | - | 5 | 412 | |||
AK134185.1 | - | 3 | 142 | |||
AK141682.1 | - | 3 | 189 | |||
AK147617.1 | - | 5 | 511 | |||
AK163964.1 | - | 3 | 132 | |||
AY027917.1 | - | 3 | 167 | |||
BC041773.1 | - | 2 | 116 | 3986207 | ||
CJ124920.1 | - | 3 | 327 |