Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ropn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030744.2 | NP_109669.1 | 5, 6 | 652 | ||
GenBank mRNA | AF178531.1 | - | 4, 5 | 575 | ||
AK160142.1 | - | 3, 4 | 394 | |||
BC049544.1 | - | 5, 6 | 652 | 6740976 |