Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030889.2 | NP_112151.2 | 6, 7 | 1054 | ||
GenBank mRNA | AB093294.1 | - | 6, 7 | 1039 | ||
AK053853.1 | - | 6, 7 | 1024 | |||
AY004316.1 | - | 6, 7 | 1023 | |||
BC138873.1 | - | 6, 7 | 980 | 8861895 | ||
BC138875.1 | - | 6, 7 | 980 | 8861897 |