Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167763.1 | NP_001161235.1 | 29 | 3764 | ||
NM_031177.4 | NP_112454.2 | 29 | 3767 | |||
GenBank mRNA | AF296075.1 | - | 6 | 694 | ||
AK031308.1 | - | 29 | 3652 | |||
AK044052.1 | - | 9 | 1475 | |||
AK044456.1 | - | 29 | 3767 | |||
AK077104.1 | - | 16 | 2227 | |||
AK131115.1 | - | 29 | 3691 | |||
AK167330.1 | - | 29 | 3719 | |||
AK167412.1 | - | 29 | 3719 | |||
AK169642.1 | - | 8 | 1023 | |||
BC038826.1 | - | 13 | 3979 | |||
BC060621.1 | - | 30 | 4150 | |||
BC066083.1 | - | 29 | 3683 | 30355922 | ||
BC067415.1 | - | 29 | 3680 | 30063124 |