Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031179.2 | NP_112456.2 | 4 | 438 | ||
GenBank mRNA | AB037890.1 | - | 4 | 463 | ||
AK012632.1 | - | 4 | 397 | |||
AK140982.1 | - | 4 | 356 | |||
AK146989.1 | - | 4 | 347 | |||
AK165992.1 | - | 4 | 442 | |||
AK171454.1 | - | 4 | 356 | |||
BC003419.1 | - | 4 | 341 | |||
BC028959.1 | - | 4 | 341 | |||
BC037098.1 | - | 4 | 425 | |||
BC058563.1 | - | 4 | 341 | |||
BC060125.1 | - | 4 | 334 | |||
BC080754.1 | - | 4 | 357 | |||
CK331410.1 | - | 4 | 438 |