Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031376.3 | NP_113553.1 | 5 | 885 | ||
GenBank mRNA | AF293806.1 | - | 5 | 885 | ||
AK087722.1 | - | 3 | 354 | |||
AK089785.1 | - | 5 | 918 | |||
AK150114.1 | - | 3 | 354 | |||
AK153548.1 | - | 3 | 336 | |||
AK154326.1 | - | 5 | 913 | |||
AK170130.1 | - | 5 | 918 | |||
AK170925.1 | - | 5 | 918 | |||
AK171021.1 | - | 5 | 913 | |||
AK172670.1 | - | 2 | 1337 | |||
BC113141.1 | - | 5 | 844 | 40047744 |